5-Methoxy-2-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A106705-10G A106705-1G A106705-25G A106705-5G
77025-478EA 37 USD
77025-478 77025-480 77025-482 77025-484
5-Methoxy-2-nitrobenzonitrile 95%
5-Methoxy-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MDL Number: MFCD09056823
CAS Number: 38469-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR