Hexaethylene glycol monobenzyl ether 98%

Supplier: Ambeed
Warning

A106604-100MG A106604-1G A106604-250MG A106604-5G
77008-624EA 9 USD
77008-624 77008-626 77008-628 77008-630
Hexaethylene glycol monobenzyl ether 98%
Hexaethylene glycol monobenzyl ether

Precaución: for Research Use Only

Formula: C₁₉H₃₂O₇
MDL Number: MFCD06797107
CAS Number: 24342-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR