1-(tert-Butoxycarbonyl)-5-oxopiperazine-2-carboxylic acid 95%

Supplier: Ambeed
Danger

A106515-100MG A106515-250MG A106515-5G A106515-1G
76982-446EA 46.68 USD
76982-446 76982-448 77570-388 77682-979
1-(tert-Butoxycarbonyl)-5-oxopiperazine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-5-oxopiperazine-2-carboxylic acid

Precaución: For research use only.

Formula: C₁₀H₁₆N₂O₅
MW: 244.24 g/mol
MDL Number: MFCD10698230
CAS Number: 1246553-28-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR