5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine 95%

Supplier: Ambeed
Warning

A106344-1G A106344-250MG
77025-458EA 110.34 USD
77025-458 77025-460
5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine 95%
5-(4-Bromo-2-fluorophenyl)-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₅BrFN₃S
MW: 274.11 g/mol
MDL Number: MFCD09999212
CAS Number: 299937-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR