tert-Butyl (3-aminocyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

A106279-100MG A106279-1G A106279-250MG A106279-5G
76994-944EA 46.68 USD
76994-944 76994-946 76994-948 76994-950
tert-Butyl (3-aminocyclobutyl)carbamate 97%
tert-Butyl (3-aminocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
MDL Number: MFCD12755997
CAS Number: 1090904-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR