1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde 97%
Supplier: Ambeed
|
Warning
|
A106250-100MG
A106250-1G
A106250-250MG
A106250-5G
76994-936EA
8.49
USD
76994-936
76994-938
76994-940
76994-942
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde 97%
1H-Pyrrolo[3,2-c]pyridine-3-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆N₂O MW: 146.15 g/mol |
MDL Number:
MFCD08690128 CAS Number: 933717-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...