4-Amino-1-methyl-4H-1,2,4-triazol-1-ium iodide 97%

Supplier: Ambeed
Warning

A106213-10G A106213-1G
76982-432EA 48.81 USD
76982-432 76982-434
4-Amino-1-methyl-4H-1,2,4-triazol-1-ium iodide 97%
4-Amino-1-methyl-4H-1,2,4-triazol-1-ium iodide

Precaución: for Research Use Only

Formula: C₃H₇IN₄
MW: 226.02 g/mol
MDL Number: MFCD22495144
CAS Number: 39602-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR