Ethyl-4-amino-3-fluorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-amino-3-fluorobenzoate

A106172-100MG A106172-10G A106172-1G A106172-250MG A106172-25G A106172-5G
77025-440EA 5 USD
77025-440 77025-442 77025-444 77025-446 77025-448 77025-450
Ethyl-4-amino-3-fluorobenzoate 97%
Ethyl 4-amino-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD07782043
CAS Number: 73792-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR