P 22077 98%

Supplier: Ambeed
Danger

Synonyms: 1-(5-((2,4-Difluorophenyl)thio)-4-nitrothiophen-2-yl)ethanone

A106163-10MG A106163-1MG A106163-50MG A106163-5MG
76982-416EA 70.02 USD
76982-416 76982-418 76982-420 76982-422
P 22077 98%
P 22077

Precaución: for Research Use Only

Formula: C₁₂H₇F₂NO₃S₂
MW: 315.32 g/mol
MDL Number: MFCD22580421
CAS Number: 1247819-59-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR