Azetidin-3-ylmethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminomethyl-azetidine dihydrochloride

A106141-100MG A106141-1G A106141-250MG A106141-5G
77025-430EA 16.98 USD
77025-430 77025-432 77025-434 77025-436
Azetidin-3-ylmethanamine dihydrochloride 97%
Azetidin-3-ylmethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₂
MW: 159.0600 g/mol
MDL Number: MFCD10565803
CAS Number: 221095-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR