5-Bromo-2-isopropoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A106118-10G A106118-250MG
77025-426EA 647.15 USD
77025-426 77025-428
5-Bromo-2-isopropoxybenzaldehyde 97%
5-Bromo-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD02257437
CAS Number: 138505-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR