2-(Chloromethyl)-4-methylquinazoline 98%

Supplier: Ambeed
Danger

A106108-1G A106108-25G A106108-5G A106108-10G
77025-420EA 6.37 USD
77025-420 77025-422 77025-424 77670-520
2-(Chloromethyl)-4-methylquinazoline 98%
2-(Chloromethyl)-4-methylquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂
MW: 192.65 g/mol
MDL Number: MFCD09807547
CAS Number: 109113-72-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR