2-Cyano-2-methylpropanoic acid 98%

Supplier: Ambeed
Danger

A105950-100MG A105950-1G A105950-250MG A105950-5G A105950-25G
76994-926EA 9 USD
76994-926 76994-928 76994-930 76994-932 77880-862
2-Cyano-2-methylpropanoic acid 98%
2-Cyano-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MDL Number: MFCD11100210
CAS Number: 22426-30-8
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR