Methyl 2-(bromomethyl)-5-cyanobenzoate 95%

Supplier: Ambeed
Danger

A105946-100MG A105946-1G
77008-600EA 41 USD
77008-600 77008-602
Methyl 2-(bromomethyl)-5-cyanobenzoate 95%
Methyl 2-(bromomethyl)-5-cyanobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MDL Number: MFCD18251143
CAS Number: 421551-82-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR