7-Methoxy-7-oxoheptanoic acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A105938-1G A105938-25G A105938-5G A105938-250MG A105938-100G
76982-410EA 23 USD
76982-410 76982-412 76982-414 77682-975 77889-737
7-Methoxy-7-oxoheptanoic acid 95%, Ambeed.Inc
7-Methoxy-7-oxoheptanoic acid

Precaución: For research use only.

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
MDL Number: MFCD00040445
CAS Number: 20291-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR