4-Sulfamoylbenzoic acid 98%

Supplier: Ambeed
Warning

A105921-100G A105921-25G A105921-1000G
76994-922EA 42 USD
76994-922 76994-924 77880-861
4-Sulfamoylbenzoic acid 98%
4-Sulphamoylbenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₄S
MDL Number: MFCD00007938
CAS Number: 138-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR