3-Bromo-5-fluoroisonicotinamide 97%
Supplier: Ambeed
|
Warning
|
A105911-100MG
A105911-1G
A105911-250MG
76982-400EA
41
USD
76982-400
76982-402
76982-404
3-Bromo-5-fluoroisonicotinamide 97%
3-Bromo-5-fluoroisonicotinamide
Precaución: for Research Use Only
Formula:
C₆H₄BrFN₂O MW: 219.011 g/mol |
MDL Number:
MFCD20524845 CAS Number: 1353636-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...