Fmoc-Asp(OAll)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asp(allyl ester)-OH, N-Fmoc-L-aspartic acid 4-allyl ester

A105894-100G A105894-10G A105894-1G A105894-25G A105894-5G A105894-500G
76994-912EA 287 USD
76994-912 76994-914 76994-916 76994-918 76994-920 77880-860
Fmoc-Asp(OAll)-OH 97%
Fmoc-Asp(OAll)-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₁NO₆
MW: 395.41 g/mol
MDL Number: MFCD00190874
CAS Number: 146982-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR