4-(Piperidin-4-yloxy)benzamide hydrochloride 95%

Supplier: Ambeed
Warning

A105851-100MG A105851-250MG
76982-394EA 20 USD
76982-394 76982-396
4-(Piperidin-4-yloxy)benzamide hydrochloride 95%
4-(Piperidin-4-yloxy)benzamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD26383916
CAS Number: 857048-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR