Boc-2,4-dichloro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid

A105815-1G A105815-5G
76994-908EA 41.38 USD
76994-908 76994-910
Boc-2,4-dichloro-L-phenylalanine 97%
Boc-2,4-dichloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.2 g/mol
MDL Number: MFCD01862942
CAS Number: 114873-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR