2,4-Dichloro-5-fluoroquinazoline 97%

Supplier: Ambeed
Warning

A105726-1G A105726-5G
77025-412EA 92.3 USD
77025-412 77025-414
2,4-Dichloro-5-fluoroquinazoline 97%
2,4-Dichloro-5-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₃Cl₂FN₂
MW: 217.0272 g/mol
MDL Number: MFCD09954883
CAS Number: 87611-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR