1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-(methylamino)piperidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Balofloxacin

A105718-1G A105718-1MG A105718-250MG A105718-25MG A105718-5G
76994-898EA 153.84 USD
76994-898 76994-900 76994-902 76994-904 76994-906
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-(methylamino)piperidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-(methylamino)piperidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₃O₄
MW: 389.42 g/mol
MDL Number: MFCD00864925
CAS Number: 127294-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR