3,3',5,5'-Tetrabromo-2,2'-bithiophene 98%
Supplier: Ambeed
|
Warning
|
A105667-1G
A105667-25G
A105667-5G
A105667-100G
77025-404EA
7
USD
77025-404
77025-406
77025-408
77025-410
77666-546
3,3',5,5'-Tetrabromo-2,2'-bithiophene 98%
3,3',5,5'-Tetrabromo-2,2'-bithiophene
Precaución: for Research Use Only
Formula:
C₈H₂Br₄S₂ |
MDL Number:
MFCD00114807 CAS Number: 125143-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...