1-Cyclopropylethylamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropylethanamine hydrochloride

A105636-100MG A105636-1G A105636-250MG A105636-5G A105636-25G
77025-390EA 6 USD
77025-390 77025-392 77025-394 77599-546 77871-406
1-Cyclopropylethylamine hydrochloride 95%
1-Cyclopropylethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN
MDL Number: MFCD06685893
CAS Number: 42390-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR