Methyl-4-amino-3-methylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-3-methylbenzoate

A105548-100G A105548-10G A105548-25G A105548-500G A105548-5G
76994-880EA 37.14 USD
76994-880 76994-882 76994-884 76994-886 76994-888
Methyl-4-amino-3-methylbenzoate 97%
Methyl 4-amino-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00102230
CAS Number: 18595-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR