2-Isopropyl-2H-pyrazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 1-Isopropyl-1H-pyrazole-5-carbaldehyde

A105540-100MG A105540-1G A105540-250MG A105540-5G
76994-872EA 38.2 USD
76994-872 76994-874 76994-876 76994-878
2-Isopropyl-2H-pyrazole-3-carbaldehyde 95%
2-Isopropyl-2H-pyrazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD03130042
CAS Number: 100305-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR