5-Bromo-6-methoxypyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methoxynicotinic acid

A105429-100MG A105429-1G A105429-250MG A105429-5G
77008-574EA 8 USD
77008-574 77008-576 77008-578 77008-580
5-Bromo-6-methoxypyridine-3-carboxylic acid 97%
5-Bromo-6-methoxypyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD11226508
CAS Number: 1186194-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR