5-Bromo-4-methylnicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-methylpyridine-3-carbonitrile

A105369-100MG A105369-250MG A105369-1G
77008-570EA 111.4 USD
77008-570 77008-572 77610-826
5-Bromo-4-methylnicotinonitrile 95%
5-Bromo-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD07439978
CAS Number: 890092-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR