Fmoc-threoninol 97%
Supplier: Ambeed
|
Warning
|
A105209-1G
A105209-250MG
A105209-25G
A105209-5G
A105209-100G
A105209-10G
76994-838EA
11
USD
76994-838
76994-840
76994-842
76994-844
77880-852
77880-853
Fmoc-threoninol 97%
Fmoc-threoninol
Formula:
C₁₉H₂₁NO₄ |
MDL Number:
MFCD00270218 CAS Number: 176380-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...