3-(4-(Benzyloxy)-3-methoxyphenyl)propan-1-ol 97%

Supplier: Ambeed
Warning

A105197-1G A105197-250MG
77008-560EA 53.05 USD
77008-560 77008-562
3-(4-(Benzyloxy)-3-methoxyphenyl)propan-1-ol 97%
3-(4-(Benzyloxy)-3-methoxyphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₃
MW: 272.34 g/mol
MDL Number: MFCD09926773
CAS Number: 57371-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR