4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-piperazine-benzoic acid

A105156-10G A105156-1G A105156-250MG A105156-25G A105156-5G A105156-100G
77008-550EA 66.84 USD
77008-550 77008-552 77008-554 77008-556 77008-558 77682-967
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid 95%
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid

Precaución: For research use only.

Formula: C₁₆H₂₂N₂O₄
MW: 306.3569 g/mol
MDL Number: MFCD04115067
CAS Number: 162046-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR