CAPS (3-(cyclohexylamino)propanesulfonic acid) 96%

Supplier: Ambeed
Warning

Synonyms: 3-Cyclohexylaminopropanesulphonic acid

A105150-1000G A105150-100G A105150-10G A105150-25G A105150-500G
76994-814EA 98.67 USD
76994-814 76994-816 76994-818 76994-820 76994-822
CAPS (3-(cyclohexylamino)propanesulfonic acid) 96%
CAPS (3-(cyclohexylamino)propanesulphonic acid)

Precaución: for Research Use Only

Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
MDL Number: MFCD00003837
CAS Number: 1135-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR