(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-amino-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate 98%

Supplier: Ambeed
Warning

A104957-100G A104957-250MG A104957-5G
77025-378EA 72 USD
77025-378 77025-380 77025-382
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-amino-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate 98%
(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(6-amino-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₅O₇
MDL Number: MFCD00057001
CAS Number: 7387-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR