(R)-3-(Boc-amino)-3-phenylpropionic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, (R)-N-BOC-3-Amino-3-phenylpropanoic acid
A104914-100MG
A104914-1G
A104914-250MG
A104914-25G
A104914-5G
76994-798EA
13.8
USD
76994-798
76994-800
76994-802
76994-804
76994-806
(R)-3-(Boc-amino)-3-phenylpropionic acid 97%
(R)-3-(Boc-amino)-3-phenylpropionic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol |
MDL Number:
MFCD01320859 CAS Number: 161024-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...