5-Bromo-2-fluoro-3-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluoro-3-picoline

A104894-100G A104894-10G A104894-1G A104894-25G A104894-5G
76994-778EA 124 USD
76994-778 76994-780 76994-782 76994-784 76994-786
5-Bromo-2-fluoro-3-methylpyridine 97%
5-Bromo-2-fluoro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01
MDL Number: MFCD03095091
CAS Number: 29312-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR