2,4-Dimethoxypyridine 98%

Supplier: Ambeed
Warning

A104819-100G A104819-10G A104819-1G A104819-5G A104819-25G
77008-534EA 195 USD
77008-534 77008-536 77008-538 77008-540 77889-733
2,4-Dimethoxypyridine 98%
2,4-Dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD01646187
CAS Number: 18677-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR