1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oic acid 98%

Supplier: Ambeed
Warning

A104811-100MG A104811-10G A104811-250MG A104811-25G A104811-1G A104811-5G
76982-360EA 19.1 USD
76982-360 76982-362 76982-364 76982-366 77309-360 77309-362
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oic acid 98%
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oic acid

Precaución: for Research Use Only

Formula: C₂₆H₃₃NO₈
MW: 487.54 g/mol
MDL Number: MFCD06656472
CAS Number: 557756-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR