4-Chloro-6-isopropoxypyrimidine 97%

Supplier: Ambeed
Warning

A104733-10G A104733-1G A104733-5G
77025-360EA 161.26 USD
77025-360 77025-362 77025-364
4-Chloro-6-isopropoxypyrimidine 97%
4-Chloro-6-isopropoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.6100 g/mol
MDL Number: MFCD11847658
CAS Number: 83774-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR