H-Lys(boc)-OMe·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-Lys(Boc)-OMe·HCl, Nepsilon-Boc-L-lysine methyl ester hydrochloride, H-Lys(Boc)-OMe.HCl

A104612-100G A104612-10G A104612-1G A104612-25G A104612-5G A104612-250MG
76994-760EA 219 USD
76994-760 76994-762 76994-764 76994-766 76994-768 77670-512
H-Lys(boc)-OMe·HCl 98%
H-Lys(boc)-OMe·HCl

Precaución: for Research Use Only

Formula: C₁₂H₂₅ClN₂O₄
MDL Number: MFCD00076959
CAS Number: 2389-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR