1-(2-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A104455-1G A104455-250MG
76982-350EA 222.79 USD
76982-350 76982-352
1-(2-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carboxylic acid 98%
1-(2-(Trifluoromethyl)phenyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₇F₃N₂O₂
MW: 256.18 g/mol
MDL Number: MFCD15527664
CAS Number: 1260855-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR