1-(3-Hydroxy-1-azetidinyl)-ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Hydroxyazetidin-1-yl)ethanone

A104320-1G A104320-250MG A104320-5G
77025-348EA 74.27 USD
77025-348 77025-350 77666-510
1-(3-Hydroxy-1-azetidinyl)-ethanone 98%
1-(3-Hydroxy-1-azetidinyl)-ethanone

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD07366667
CAS Number: 118972-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR