4-(4-Biphenylyl)butyric acid 97%

Supplier: Ambeed
Warning

A104296-100MG A104296-1G A104296-250MG A104296-5G
77025-340EA 7 USD
77025-340 77025-342 77025-344 77025-346
4-(4-Biphenylyl)butyric acid 97%
4-(4-Biphenylyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₂
MW: 240.30 g/mol
MDL Number: MFCD03427545
CAS Number: 6057-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR