TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98%

Supplier: Ambeed
Warning

A104132-1000G A104132-25G A104132-500G A104132-100G
77025-334EA 82 USD
77025-334 77025-336 77025-338 77592-100 77599-522
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98%
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)

Precaución: for Research Use Only

Formula: NH₂C(CH₂OH)₃·HCl
MDL Number: MFCD00012590
CAS Number: 1185-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR