(3R,4S)-4-(4-(Benzyloxy)phenyl)-1-(4-fluorophenyl)-3-(3-(4-fluorophenyl)-3-oxopropyl)azetidin-2-one 98%

Supplier: Ambeed
Warning

A104063-1G A104063-5G
77025-330EA 22 USD
77025-330 77025-332
(3R,4S)-4-(4-(Benzyloxy)phenyl)-1-(4-fluorophenyl)-3-(3-(4-fluorophenyl)-3-oxopropyl)azetidin-2-one 98%
(3R,4S)-4-(4-(Benzyloxy)phenyl)-1-(4-fluorophenyl)-3-(3-(4-fluorophenyl)-3-oxopropyl)azetidin-2-one

Precaución: for Research Use Only

Formula: C₃₁H₂₅F₂NO₃
MDL Number: MFCD13185976
CAS Number: 190595-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR