2-Amino-3-methylpyridine 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

A104003-25G A104003-100G
76994-708EA 6.37 USD
76994-708 77599-514
2-Amino-3-methylpyridine 97%
2-Amino-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00006320
CAS Number: 1603-40-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR