3-Bromo-2,4-difluoronitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1,3-difluoro-4-nitrobenzene

A103998-100MG A103998-1G A103998-250MG A103998-25G A103998-5G
77025-324EA 12.74 USD
77025-324 77025-326 77025-328 77565-806 77570-236
3-Bromo-2,4-difluoronitrobenzene 98%
3-Bromo-2,4-difluoronitrobenzene

Precaución: For research use only.

Formula: C₆H₂BrF₂NO₂
MW: 237.986 g/mol
MDL Number: MFCD12545882
CAS Number: 103977-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR