Boc-L-3-benzothienylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-β-(3-benzothienyl)-Ala-OH, (S)-3-(Benzo[b]thiophen-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

A103940-1G A103940-250MG A103940-5G
76994-702EA 56.23 USD
76994-702 76994-704 76994-706
Boc-L-3-benzothienylalanine 95%
Boc-L-3-benzothienylalanine

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₄S
MW: 321.391 g/mol
MDL Number: MFCD00237538
CAS Number: 154902-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR