2-Acetyl-5-methylfuran 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

A103878-100G A103878-10G A103878-25G A103878-500G A103878-5G
76994-692EA 33.95 USD
76994-692 76994-694 76994-696 76994-698 76994-700
2-Acetyl-5-methylfuran 98%
2-Acetyl-5-methylfuran

Precaución: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00003243
CAS Number: 1193-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR