(E)-N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-methylacetamide 95%

Supplier: Ambeed
Warning

A103844-100MG A103844-1G
77008-514EA 20.16 USD
77008-514 77008-516
(E)-N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-methylacetamide 95%
(E)-N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-methylacetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₂
MW: 246.31 g/mol
MDL Number: MFCD09833226
CAS Number: 1227694-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR