Diethyl-3-oxoglutarate 95%

Supplier: Ambeed
Warning

Synonyms: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

A103827-1000G A103827-100G A103827-25G A103827-500G A103827-5G
76994-674EA 187.78 USD
76994-674 76994-676 76994-678 76994-680 76994-682
Diethyl-3-oxoglutarate 95%
Diethyl-3-oxoglutarate

Precaución: for Research Use Only

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
MDL Number: MFCD00009200
CAS Number: 105-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR